Bacitracin zinc salt, potency min. 60 Units/mg
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1405-89-6
:
Formula:C₆₆H₁₀₁N₁₇O₁₆SZn
MDL Number:MFCD28168035
Melting Pt:250 °C (dec.)
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to yellow to beige to light grey Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5.0 %
- Potency:≥60 U/mg (On dried substance)
- pH:6.0 to 7.5 (10 % solution)
- Zinc (Zn):4.0 to 6.0 % (on dried substance)
Specification Test Results
Frequently Bought Together