Cresol red sodium salt 0,04% (w/v) in aqueous solution
Catalog # 42752.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:404.42 g/mol
Storage Temperature:Ambient
EINECS:263-654-8
CAS Number:62625-29-0
Formula:C₂₁H₁₇NaO₅S
MDL Number:MFCD00001618