Order Entry
Northern Ireland
ContactUsLinkComponent
 
2-Deoxy-D-glucose, Millipore®
Catalog # 25972-1
CAS Number:  
2-Deoxy-D-glucose, Millipore®
Catalog # 25972-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.16 g/mol
EINECS:205-823-0
CAS Number:154-17-6
 :MFCD00007013
Formula:C₆H₁₂O₅
MDL Number:MFCD00151328