Order Entry
Northern Ireland
ContactUsLinkComponent
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine ≥97%
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine ≥97%
Catalog # H31993.03
CAS Number:  
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine ≥97%
Catalog # H31993.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.99 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:89402-29-9
Formula:C₆H₂BrF₄N
MDL Number:MFCD10699118