

Zirconium(IV) ethoxide 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18267-08-8
:
Formula:C₈H₂₀O₄Zr
MDL Number:MFCD00015654
:271.47 g/mol
Boiling Pt:235 °C (7mbar)
Melting Pt:200 °C
Storage Temperature:Ambient
UN:3181
ADR:4.1,II
:GHS02
:GHS07
Specification Test Results
- Appearance:White Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Assay:≥98.5 %
Specification Test Results
Frequently Bought Together