PIG® fibreglass chemical trays
: NEW PIG
Catch drips and control messes with these smooth, seamless fibreglass trays.
- Reinforced thermoset composite fibreglass resists chemicals and retains its shape, even under heavy loads
- Smooth corners and seamless design allows trays to be steam cleaned or sterilized for repeated use
- Molded edges safely contain minor spills or leaks from containers or oily parts
Chemical-resistant trays are easy to clean.
Frequently Bought Together