Automatic saccharimeters, ADS420
Supplier: BELLINGHAM STANLEY
Certificates
About this item
Automatic saccharimeter designed for sugar purity in laboratories and tare houses.
- PC software for 'purity' calculation
- LED light source that requires no maintenance
- Suitable for the measurement of samples with low transmittance
A single temperature sensor provides the measurement for sugar compensation whilst quartz compensation facilitates accurate verification and calibration using a quartz control plate by accessing the span facility in the setup menu.
These saccharimeters can measure dark samples of up to 3 OD, for optimum performance it is ideal to remove colour and suspended solids to provide a clear solution or one of less than 2,0 OD.
Certifications: ICUMSA
Frequently Bought Together