
Diphosphoryl tetrachloride
Catalog # 22362.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:251.76 g/mol
Boiling Pt:101 °C (10 mmHg)
Melting Pt:–17 °C
Density:1,82
Storage Temperature:Ambient
EINECS:236-824-4
UN:3264
ADR:8,II
:GHS05
CAS Number:13498-14-1
Formula:Cl₄O₃P₂
MDL Number:MFCD00061542