Betaine hydrochloride
Catalog # PROOMM0611.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:153.61 g/mol
Storage Temperature:Ambient
EINECS:209-683-1
CAS Number:590-46-5
Formula:C₅H₁₂ClNO₂
MDL Number:MFCD00011903