Polyethylene glycol dimethylether, M.W. 250 for synthesis, Sigma-Aldrich®
Catalog # 8.14173.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 ml
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-23 °C
- Molar Mass:250 g/mol
- Boiling Point:250 °C
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:137 °C
- Refractive Index:1.439 (20 °C, 589 nm)
- Explosion Limit:0.5 %(V)
- Density:1.03 g/cm3 (20 °C)
- pH:(100 g/l, H2O, 20 °C) neutral
- Viscosity kinematic:7.8 mm2/s (20 °C)
Specification Test Results
About this item
Synonyms: Poly(ethylene glycol) dimethyl ether Mn 250
:
CAS Number:24991-55-7
Formula:H₃C(OCH₂CH₂)nOCH₃
MDL Number:MFCD00147972
:M-Clarity™ quality level = MQ200