Dragendorff's reagent derivatisation reagent, for TLC, Supelco®
Catalog # 44578-100ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
Boiling Pt:77 °C (1013 hPa)
Density:1,2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:–4 °C
Storage Temperature:Ambient
CAS Number:39775-75-2
MDL Number:MFCD00283106