


Hygromycin B, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:31282-04-9
:
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479
:527.5 g/mol
Storage Temperature:Refrigerator
EINECS:250-545-5
UN:3462
ADR:6.1,I
:GHS06
:GHS05
:GHS08
Frequently Bought Together