
S-Methylisothiouronium sulphate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:867-44-7
:
Formula:C₄H₁₂N₄S₂·H₂SO₄
MDL Number:MFCD00013055
:278.38 g/mol
Melting Pt:∼ 235 °C (dec.)
Density:1,28
EINECS:212-759-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together