ContactUsLinkComponent
S-Methylisothiouronium sulphate 98%
Irritant
S-Methylisothiouronium sulphate 98%
Certificates
undefined
Irritant
S-Methylisothiouronium sulphate 98%
Specification Test Results
  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥97.5 %
  • HPLC:
    ≥97.5 %