Vitamin K1 (phytomenadione, phylloquinone)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:84-80-0
:81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063
:450.6958 g/mol
Merck Index:14,7380
Boiling Pt:∼140 °C (0,001 m)
Melting Pt:∼–20 °C
Density:0,984
Flash Pt:>110 °C (230 °F)
EINECS:201-564-2
Related Information
Frequently Bought Together