Pesticide reference standard, trans-Permethrin 10 µg/ml in cyclohexane, Supelco®
Catalog # 36893-2ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- CAS No.:61949-77-7
- Description:trans-Permethrin 10 µg/ml in cyclohexane
- M-Clarity quality level:MQ100
- Formula:C₂₁H₂₀C₁₂O₃
- Description:trans-Permethri