Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
γ-Cyclodextrine hydrate

γ-Cyclodextrine hydrate

γ-Cyclodextrine hydrate
Certificates
γ-Cyclodextrine hydrate

2 Options Available Below

Product Details & Documents

CAS No.: 91464-90-3 Molecular Formula: C48H80O40.xH2O Formula Weight: 1297.15(anhy) Melting Point: ca 267° dec. Beilstein Reference: 5725162 Merck Reference: 14,2718 Optical Rotation: +177° (c=1 in water) on dry basis MDL No.: MFCD00149574

Synonyms:  γ-Cyclodextrin hydrate, γ-Cyclodextrin xhydrate
  • CAS Number:91464-90-3
  • Formula:C₄₈H₈₀O₄₀·H₂O
  • MDL Number:MFCD00149574
  • Molecular Weight:1297.15 g/mol (anhy)
  • Merck Index:14,2718
  • Melting Pt:∼ 267 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:241-482-4
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - £41.20
    £41.20
    1 * 100 mg
       £41.20

    Specifications:
    More
    Specifications
    Cat. No.
    L11271.MC
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - £126.00
    £126.00
    1 * 500 mg
       £126.00

    Specifications:
    More
    Specifications
    Cat. No.
    L11271.ME
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.