Order Entry
Northern Ireland
ContactUsLinkComponent
 
(2-Hydroxylpropyl)-β-cyclodextrine
Catalog # A0367.0100
Supplier:  PanReac AppliChem
CAS Number:  
(2-Hydroxylpropyl)-β-cyclodextrine
Catalog # A0367.0100
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:128446-35-5
 :MFCD00069372
Formula:C₆₃H₁₁₂O₄₂
MDL Number:MFCD16621721