3,6-Dimethyl-1,4-dioxane-2,5-dione 99%
:95-96-5
:
:C₆H₈O₄
:MFCD00011685
:138…142 °C (8 mmHg)
:122…126 °C
:144.13 g/mol
:Refrigerator
:202-468-3
:GHS07
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:122 to 126 °C
- GC:≥98.5 %
Frequently Bought Together