

Calcium cyanamide, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:156-62-7
:
Formula:CCaN₂
MDL Number:MFCD00064894
:80.11 g/mol
Merck Index:14,1662
Melting Pt:1340 °C
Density:2,29
Storage Temperature:Ambient
EINECS:205-861-8
UN:1403
ADR:4.3,III
:GHS05
:GHS07
Related Information
Frequently Bought Together