Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
(R)-(-)-Prolinol

(R)-(-)-Prolinol

Glass bottle for solids, 5 g
Catalog # MOLE45777293-5G
Marketsource
Supplier:   Molekula
(R)-(-)-Prolinol
Catalog # MOLE45777293-5G
Supplier:  Molekula
Marketsource
CAS Number:  68832-13-3
(R)-(-)-Prolinol

Glass bottle for solids, 5 g

CAS Number:  68832-13-3
Price
£74.00
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  D-Prolinol, (2R)-pyrrolidin-2-ylmethanol, D(-)-Prolinol
  • CAS Number:68832-13-3
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00064321
  • Molecular Weight:101.1 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.