Sulphacetamide ≥98%
Catalog # A19836.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:144-80-9
:
Formula:C₈H₁₀N₂O₃S
MDL Number:MFCD00066501
:214.24 g/mol
Merck Index:14,8899
Melting Pt:182…184 °C
EINECS:205-640-6
:GHS08
Storage Temperature:Ambient