Order Entry
Northern Ireland
ContactUsLinkComponent
Manganese(II) perchlorate hexahydrate 99%
Oxidizing
Manganese(II) perchlorate hexahydrate 99%
Catalog # ACRO316511000
CAS Number:  
undefined
Oxidizing
Manganese(II) perchlorate hexahydrate 99%
Catalog # ACRO316511000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Pink Crystalline solid
  • Assay:
    ≥98.5 %

Specification Test Results

About this item

 :
 :361.931 g/mol
Melting Pt:165 °C
Storage Temperature:Ambient
EINECS:237-390-9
UN:1481
ADR:5.1,II
 :GHS03
CAS Number:15364-94-0
Formula:Cl₂MnO₈·6H₂O
MDL Number:MFCD00150255