Triethyl phosphonoformiate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1474-78-8
:
Formula:C₇H₁₅O₅P
MDL Number:MFCD00009108
:210.166 g/mol
:M-Clarity™ quality level = MQ200
EINECS:216-016-8
Frequently Bought Together