
Ethylene glycol dimethacrylate (ethylene dimethacrylate) stabilised for synthesis, Sigma-Aldrich®
: Merck
:97-90-5
:
:C₁₀H₁₄O₄
:MFCD00008590
:198.21 g/mol
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
:202-617-2
:GHS07
:Ambient
- Solubility:- 5 g/l (20 °C)
- Melting Point:-20 °C
- Molar Mass:198.22 g/mol
- Boiling Point:240 °C (1013 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:101.5 °C
- Density:1.05 g/cm3 (20 °C)
Frequently Bought Together