Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, Fluorene 100 µg/ml in acetonitrile
Organic reference standard, Fluorene 100 µg/ml in acetonitrile
Organic reference standard, Fluorene 100 µg/ml in acetonitrile
PROONE5047
:
LGC Standards PROMOCHEM
:
Organic reference standard, Fluorene 100 µg/ml in acetonitrile
PROONE5047
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,5 mL
CAS No.:
86-73-7
Description:
Fluorene 100 µg/ml in acetonitrile
Formula:
C₁₃H₁₀
Description:
Fluorene