Order Entry
Northern Ireland
ContactUsLinkComponent
Bis(2-chloroethyl)ammonium chloride for synthesis, Sigma-Aldrich®
Bis(2-chloroethyl)ammonium chloride for synthesis, Sigma-Aldrich®
Catalog # 8.20159.0100
Supplier:  Merck
CAS Number:  
Bis(2-chloroethyl)ammonium chloride for synthesis, Sigma-Aldrich®
Catalog # 8.20159.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    100 g/l (20 °C)
  • Melting Point:
    212 - 215 °C (decomposition)
  • Molar Mass:
    178.48 g/mol

Specification Test Results

About this item

 :
 :178.49 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:212-479-5
UN:1759
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:821-48-7
Formula:C₄H₁₀Cl₃N
MDL Number:MFCD00012515