Order Entry
Northern Ireland
ContactUsLinkComponent
Cacodylic acid sodium salt trihydrate for synthesis, Sigma-Aldrich®
Toxic
Environment
Cacodylic acid sodium salt trihydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Environment
Cacodylic acid sodium salt trihydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    2000 g/l (20 °C)
  • Melting Point:
    60 °C
  • Molar Mass:
    214.03 g/mol
  • Bulk Density:
    870 kg/m3
  • pH:
    9.0 - 9.5 (100 g/l, H2O, 20 °C)
  • Synonyms:
    Dimethylarsinic acid sodium salt, Cacodylic acid sodium salt
  • Hill Formula:
    C2H6AsNaO2 * 3 H2O

Specification Test Results

Related Information

Frequently Bought Together