N-(2-Pyridyl)thiourea ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14294-11-2
:
Formula:C₆H₇N₃S
MDL Number:MFCD00041227
:153.21 g/mol
Melting Pt:146…149 °C
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together