Sarcosine ethyl ester hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52605-49-9
:
Formula:C₅H₁₁NO₂·HCl
MDL Number:MFCD00012506
:153.61 g/mol
Melting Pt:122…126 °C
EINECS:258-037-5
Related Information
Frequently Bought Together