



Hexafluoroacetone trihydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34202-69-2
:
Formula:C₃F₆O·3H₂O
MDL Number:MFCD00149061
:220.07 g/mol
Boiling Pt:105…106 °C
Melting Pt:18…21 °C
Density:1,579
EINECS:211-676-3
UN:2552
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together