Order Entry
Northern Ireland
ContactUsLinkComponent
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene for synthesis
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene for synthesis
Catalog # 8.14227.0005
Supplier:  Merck
CAS Number:  
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene for synthesis
Catalog # 8.14227.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 ml
  • Seq:
    2210

Specifications

About this item

 :
 :263.93 g/mol
EINECS:218-623-3
CAS Number:2207-27-4
Formula:C₇H₆Cl₄O₂
MDL Number:MFCD00001353