6-Methoxy-1-tetralone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1078-19-9
:
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00001695
Merck Index:14,6000
Boiling Pt:171 °C (11 mmHg)
Melting Pt:77…80 °C
:176.22 g/mol
Storage Temperature:Ambient
EINECS:214-078-0
Related Information
Frequently Bought Together