2(5H)-Furanone 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:497-23-4
:
Formula:C₄H₄O₂
MDL Number:MFCD00005376
Boiling Pt:212…214 °C
Melting Pt:3…4 °C
Density:1,197
Flash Pt:101 °C (213 °F)
:84.07 g/mol
EINECS:207-839-3
:GHS07
Related Information
Frequently Bought Together