


4-n-Nonylphenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-40-5
:
Formula:C₁₅H₂₄O
MDL Number:MFCD00002396
:220.35 g/mol
Boiling Pt:127…130 °C (27 mmHg)
Melting Pt:43…46 °C
Density:0,937
Flash Pt:150 °C (302 °F)
Storage Temperature:Ambient
EINECS:203-199-4
UN:2430
ADR:8,III
:GHS05
:GHS08
:GHS07
Related Information
Frequently Bought Together