Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Aldrin Solution
Supelco® Organic Reference Standard, Aldrin Solution
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Aldrin Solution
Supplier:  Merck

Specifications

  • CAS No.:
    309-00-2
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319 - H412
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00135587
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₂H₈Cl₆
  • Molecular weight:
    364,91
  • Flash point:
    6,0 °C - closed cup

Specifications

Frequently Bought Together