£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Chloroform : isoamyl alcohol
Chloroform : isoamyl alcohol (24:1) (v/v)
Chloroform : isoamyl alcohol (24:1) (v/v)
A1935.0500
:
PanReac AppliChem
:
Chloroform : isoamyl alcohol (24:1) (v/v)
A1935.0500
:
PanReac AppliChem
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
Ambient
:
1888
:
6.1,III
:
GHS08
:
GHS07
:
CHCl₃,C₅H₁₂O