Order Entry
Northern Ireland
ContactUsLinkComponent
(4-Bromophenyl)acetone 97%
(4-Bromophenyl)acetone 97%
Catalog # ACRO380220050
CAS Number:  
undefined
(4-Bromophenyl)acetone 97%
Catalog # ACRO380220050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5560 to 1.5590 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :213.07 g/mol
Storage Temperature:Ambient
CAS Number:6186-22-7
Formula:C₉H₉BrO
MDL Number:MFCD00210401