£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, (S)-(-)-Nicotine 10 µg/ml in methanol
Organic reference standard, (S)-(-)-Nicotine 10 µg/ml in methanol
PROONE9288
:
LGC Standards PROMOCHEM
:
Organic reference standard, (S)-(-)-Nicotine 10 µg/ml in methanol
PROONE9288
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
54-11-5
Description:
(S)-(-)-Nicotine 10 µg/ml in methanol
Formula:
C₁₀H₁₄N₂
Description:
(S)-(-)-Nicotine