

Acetic acid glacial, ChromAR® for TLC, Macron Fine Chemicals™
V155-06
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Assay (by GC, corrected for water):≥99.7 %
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Chloride (Cl):≤0.4 ppm
- Color (APHA):≤10
- Dilution Test:Passes Test
- Heavy Metals (by ICP-OES):≤5 ppm
- Identification:Passes Test
- Residue after Evaporation:≤5 ppm
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Sulfate (SO₄):≤0.4 ppm
- Titrable Base (meq/g):≤0.0004
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.02
- Trace Impurities - Copper (Cu):≤0.10 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Nickel (Ni):≤0.1 ppm
:
:60.05 g/mol
:118 °C (1013 hPa)
:17 °C
:1,05 g/cm³ (20 °C)
:38,5 °C
:Ambient
:200-580-7
:2789
:8,II
:01-2119475328-30
:GHS02
:GHS05
:64-19-7
:H₃CCOOH
:MFCD00036152