


2-Amino-4-bromophenol 98%
Catalog # H55763.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.02 g/mol
Melting Pt:130…135 °C
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:40925-68-6
Formula:C₆H₆BrNO
MDL Number:MFCD00235171