Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium hydroxide 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Sodium hydroxide 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # MACR6146-60
CAS Number:  
undefined
Sodium hydroxide 0.10 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # MACR6146-60
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0995 - 0.1005
  • Normality:
    0.0995 - 0.1005

Specification Test Results

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548