Order Entry
Northern Ireland
ContactUsLinkComponent
(3-Aminopropyl)triethoxysilane
Corrosive
Irritant
(3-Aminopropyl)triethoxysilane
Catalog # PIER80370
CAS Number:  
undefined
Corrosive
Irritant
(3-Aminopropyl)triethoxysilane
Catalog # PIER80370
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :221.37 g/mol
Boiling Pt:217 °C (1013 hPa)
Melting Pt:< –70 °C
Density:0,95 g/cm³ (20 °C)
Flash Pt:104 °C
EINECS:213-048-4
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:919-30-2
Formula:H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MDL Number:MFCD00008207