ContactUsLinkComponent
dNTP mix, 10 mM
dNTP mix, 10 mM
Catalog # 71004-3
CAS Number:  
dNTP mix, 10 mM
Catalog # 71004-3
CAS Number:  
Specifications
  • Pk:
    0,2 mL
  • Description:
    10 mM dNTP mix
Specifications
About this item
The 10 mM dNTP mix is a ready-to-use preparation of ultrapure dATP, dCTP, dGTP, and dTTP (monosodium salts) at a concentration of 10 mm each in sterile deionised water at pH 7,0. Qualified for any application that requires pure deoxynucleotides, such as PCR, cDNA synthesis, and fill-in reactions.
  • Qualified for enzymatic DNA synthesis
  • Free from RNase and DNase