Orders LinkContactUsLinkComponent
Cleaning fluid, 'Teepol' bleach
Cleaning fluid, 'Teepol' bleach
Catalog #
Cleaning fluid, 'Teepol' bleach
Catalog #
Supplier:  
Cleaning fluid, 'Teepol' bleach
Product Details & Documents
Solution with approximately 4% available chlorine, recommended for cleaning and disinfection.
  • Kills 99% of germs, bacteria, fungi and viruses
  • Effective against HIV and Hepatitis A
 : Do not use with acids as this will rapidly release a toxic gas.