Büchner funnels, two piece, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
PP, white.
- Autoclavable
- Robust and lightweight
- Top and bottom can be separated for easy cleaning
For filter paper or membrane filtration discs.
Frequently Bought Together