Biocide, Klercide™ Amine, sterile
Catalog # 141-1157
Supplier: ECOLAB CONTAMINATION CONTROL
Specifications
- Description:Klercide™ Amine, sterile, capped
- Packed:5 litre container
- Pk:4
Specifications
About this item
Suitable for ISO Class 5; Grade A and B (microbial count) areas; supplied sterile by gamma irradiation and double bagged in a Grade C cleanroom. Amphoteric surfactant with broad spectrum activity. Filtered to 0,2 µm.
- CEN and isolate tested
- Safe and effective: non toxic and non corrosive
- Fully compatible and ideal for a rotational regime
- SteriShield Delivery System™ ensures 6 months in-use shelf life
EN standards: EN 1276 - bactericidal activity; EN 1650 - fungicidal activity; EN 13697 bactericidal surface test.
Caution: Use biocides safely. Always read the label and product information before use.