ContactUsLinkComponent
VWR®, Spill Response Kit for Chemicals
VWR®, Spill Response Kit for Chemicals
Catalog # 121-1050
Supplier:  VWR Collection
undefined
VWR®, Spill Response Kit for Chemicals
Catalog # 121-1050
Supplier:  VWR Collection
Specifications
  • Description:
    Spill response kit for chemicals
  • Absorbency:
    10 l/kit
  • Pk:
    1 KIT
Specifications
About this item
Spill kit suitable for dangerous liquids.
  • Total absorbent capacity up to 10 litres, depending on the chemical being absorbed
  • Suitable for oils and solvents
  • Can be used for most chemicals
Supplied in a clip closed clear bag.
Delivery: Kit contains: 2× blue plastic disposal bags, 1× 6 cm×1,2m maintenance sock, 1× clip close carrier, 1× instruction sheet, 10× maintenance spill pads, 1× non branded spill kit header card, 2× plastic zip ties.