Tubing, coloured, PTFE, BOLA
Supplier: Bohlender
Certificates
About this item
PTFE, coloured.
- Temperature resistance: from −200 to +260 °C
- Universal chemical resistance
This completely imbued tubing is lightfast, different colours assure high safety against confusions. The colour pigments can possibly affect the chemical resistance.
Frequently Bought Together