(-)-Shikimic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:138-59-0
:
Formula:C₇H₁₀O₅
MDL Number:MFCD00066278
:174.15 g/mol
Merck Index:14,8480
Melting Pt:184…188 °C
Storage Temperature:Ambient
EINECS:205-334-2
Related Information
Frequently Bought Together